Acetyl-(Cys³,Nle⁴,Arg⁵,D-2-Nal⁷,Cys¹¹)-alpha-MSH (3-11) Amide Trifluoroacetate
Catalog #: H-4598.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:212370-59-7
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782253
- Protein synonyms:HS024
- Protein/peptide name:Acetyl-(Cys³,Nle⁴,Arg⁵,D-2-Nal⁷,Cys¹¹)-alpha-MSH (3-11) amide Trifluoroacetate
- Molecular weight:1266.52
- Sequence:Ac-Cys-Nle-Arg-His-D-2-Nal-Arg-Trp-Gly-Cys-NH₂
- Molecular formula:C₅₈H₇₉N₁₉O₁₀S₂
- Cat. no.:H-4598.0001BA
- Supplier No.:H-4598.0001
Specifications
About this item
The cyclic MSH analog HS024 showed about 20-fold selectivity and very high affinity (Ki = 0.29 nM) for the melanocortin-4 (MC4) receptor and also increased the food intake, indicating that blockade of the MC4 receptor is a highly effective way to increase feeding.
Customers Who Bought This Also Bought