Somatostatin-14
Catalog #: H-1490.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:38916-34-6
- Size:25 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00076762
- Protein synonyms:SRIF-14|Somatotropin Release-Inhibiting Factor|SRIF
- Protein/peptide name:Somatostatin-14
- Molecular weight:1637.90
- Sequence:H-Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys-OH
- Molecular formula:C₇₆H₁₀₄N₁₈O₁₉S₂
- Cat. no.:H-1490.0025BA
- Supplier No.:H-1490.0025
Specifications
About this item
Somatostatin-14 (SRIF) was originally isolated from ovine hypothalamic extracts on the basis of its ability to inhibit the secretion of radioimmunoassayable growth hormone by primary cultures of enzymatically dispersed rat anterior pituitary cells. SRIF is a small cyclic neuropeptide hormone with broad inhibitory effects on endocrine secretions such as the production of GH, glucagon, and insulin.
Customers Who Bought This Also Bought