C-Reactive Protein (174-185)
Catalog #: H-1344.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:160369-86-8
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00236934
- Protein synonyms:CRP
- Protein/peptide name:C-Reactive Protein (174-185)
- Molecular weight:1276.50
- Sequence:H-Ile-Tyr-Leu-Gly-Gly-Pro-Phe-Ser-Pro-Asn-Val-Leu-OH
- Molecular formula:C₆₂H₉₃N₁₃O₁₆
- Cat. no.:H-1344.0001BA
- Supplier No.:H-1344.0001
Specifications
About this item
CRP, an acute phase serum protein synthesized in response to inflammatory cytokines, produces antitumor effects in animals. The fragment CRP (174-185) shows similar activity by significantly enhancing the tumoricidal activity of human monocytes and alveolar macrophages in vitro. CRP (174-185) may therefore play a role as a biological response modifier in cancer therapy.
Customers Who Bought This Also Bought