EP Scientific, Depyrogenated Jars, with Closure, Thermo Scientific
Catalog #: EP130-02C-PF
Supplier: Thermo Scientific
Specifications
- Cap Size:53-400
- Capacity:60 ml (2 oz.)
- Ø:57 mm
- Description:Clear Wide Mouth Jar with Polypropylene Cap, PTFE Liner
- Height:48 mm
- Material:Glass
- Cat. no.:EP130-02C-PF
- Supplier No.:130-02C/PF
Specifications
About this item
Thermo Scientific™ Depyrogenated Glass Containers are specially prepared to meet endotoxin levels of less than 0.06 EU/mL for use in terminal sterilization, water sampling and product storage.
- Used for water sampling
- Cleaning and packaging is performed in a Class 100/10 Cleanroom
- Packaging and storing laboratory reagents, samples and medias
Recommended for endotoxin-free applications. Thermo Scientific also offers glass vials specially prepared to meet endotoxin levels of less than 0.06 EU/mL.
Certifications: Certificate of Analysis is included with each lot
Ordering information: Containers with additional low particle certification are available.
Customers Who Bought This Also Bought