RV5 2-STAGE,230V W/LINE CORD
Catalog #: CAR240-00-003
Supplier: Edwards
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Methanol ≥99.8%, GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Supplier: MilliporeSigma
$578.24 to $2,015.51
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80
Puritan® UniTranz-RT® Media Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
$281.52 to $1,477.95
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$156.54 to $571.97