Nitric acid 68.0 - 70.0%, GR ACS, Supelco®
Catalog #: CANX0409-2
Supplier: MilliporeSigma
CAS Number: 7697-37-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, pre-packed
- Size:2.5 L
- Supplier No.:NX0409-2
Specifications
Specification Test Results
- Aluminum (Al):0.2ppm max
- Appearance:To pass test
- Arsenic (As):0.01ppm max
- Assay:68.0-70.0%
- Calcium (Ca):0.2ppm max
- Chloride (Cl):0.5ppm max
- Chromium (Cr):0.1ppm max
- Color (APHA):10 max
- Copper (Cu):0.05ppm max
- Gold (Au):0.3ppm max
- Heavy metals (as Pb):0.2ppm max
- Iron (Fe):0.2ppm max
- Lead (Pb):0.1ppm max
- Magnesium (Mg):0.3ppm max
- Manganese (Mn):0.2ppm max
- Mercury (Hg):0.005ppm max
- Nickel (Ni):0.05ppm max
- Phosphate (PO4):0.2ppm max
- Potassium (K):0.5ppm max
- Residue after ignition:5ppm max
- Sodium (N/A):0.5ppm max
- Sulfate (SO4):1ppm max
- Tin (Sn):0.3ppm max
- Titanium (Ti):0.3ppm max
- Zinc (Zn):0.3ppm max
- Supelco®, Nitric acid 68.0 - 70.0%, GR ACS:
Specification Test Results
About this item
:63.01
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
ADR:8,
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt: 120.5 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -42 °C|Melting Pt: –42 °C
:Flash Pt:
:Flash Pt:
:Density: 1.51 g/cm³ (20 °C)
:M.W. 63.01 g/mol
Merck Index:13,06608
:Nitric acid
EINECS:231-714-2
UN:2031
:1.51
:GHS03
:GHS06
:GHS05
Storage Temperature:Ambient
:MQ300
:12,06672
Customers Who Bought This Also Bought