Order Entry
ContactUsLinkComponent
10X TE, pH 8.0 (100 mM Tris HCl, 10 mM EDTA)
Buffers, TE solutions 10X concentrates (TRIS-EDTA buffer), Rockland Immunochemicals
Catalog #: CAMB-007
CAS Number:  
Buffers, TE solutions 10X concentrates (TRIS-EDTA buffer), Rockland Immunochemicals
Catalog #: CAMB-007
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    8.0
  • Description:
    TE, 10X, DEPC Treated
  • Size:
    1 L
  • Cat. no.:
    CAMB-007
  • Supplier No.:
    MB-007

Specifications

About this item

TE buffer is a commonly used buffer solution in molecular biology, especially in procedures involving DNA, cDNA or RNA

TE buffer is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 100 mM Tris HCl and 10 mM EDTA at a pH of 8.0. Meticulously prepared using ultra pure reagents dissolved in DEPC treated molecular biology grade water.

TE is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. The purpose of TE buffer is to solubilize DNA or RNA, while protecting it from degradation.

Caution: These products ares for research use only and are not intended for therapeutic or diagnostic applications.
Customers Who Bought This Also Bought