Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis) USP, J.T.Baker®
Catalog #: CAJT3803-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7782-85-6
MOC: Highly RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, wide mouth, black T/E cap
- Size:500 g
- Supplier No.:3803-01
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Appearance (Colorless or translucent crystals or granules):Passes Test
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤ 16 ppm
- USP - Chloride (Cl):≤ 0.06 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- USP - Identification:Passes Test
- USP - Insoluble Substances:≤ 0.4 %
- USP - Loss on Drying:43.0 - 50.0 %
- USP - Sulfate (SO₄):≤ 0.2 %
- EP - Identification A:Passes Test
- EP - Identification C:Passes Test
- EP - Identification D:Passes Test
- EP - Reducing Substances:Passes Test
- EP - Monosodium Phosphate:≤ 2.5 %
- EP - Chloride (Cl):≤ 200 ppm
- EP - Sulfate (SO₄):≤ 500 ppm
- EP - Appearance of Solution:Passes Test
- EP - Iron (Fe):≤ 20 ppm
- EP - Assay (dried basis):98.0 - 101.0 %
- JP - Arsenic (As):≤ 2 ppm
- JP - Assay (Na₂HPO₄) (dried basis):98.0 - 101.0 %
- JP - Carbonate (CO₃):Passes Test
- JP - Chloride (Cl):≤ 0.014 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals:≤ 10 ppm
- JP - Identification:Passes Test
- JP - pH (1 in 50):9.0 - 9.4
- JP - Sulfate (SO₄):≤ 0.038 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- The EP results are derived from testing Sodium Phosphate, Dibasic,:
- 7-Hydrate, U.S.P. to the EP chemical specifications derived from:
- Disodium Phosphate Dihydrate and Disodium Phosphate Dodecahydrate.:
- The JP results are derived from testing Sodium Phosphate, Dibasic,:
- 7-Hydrate, U.S.P. to the JP chemical specifications derived from Dibasic:
- Sodium Phosphate Hydrate.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
About this item
Synonyms: Sodium phosphate dibasic heptahydrate,Sodium hydrogen phosphate heptahydrate
Melting Pt:48 °C
Density:1.7 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
:Management of Change (MOC) category = HR
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 48 °C
:Flash Pt:
:Flash Pt:
:M.W. 268.07 g/mol
Storage Temperature:Ambient
EINECS:231-448-7
:M.O.C. Category HR
Customers Who Bought This Also Bought