L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT2085-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 63-68-3
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:100 g
- Cat. no.:CAJT2085-5
- Supplier No.:2085-05
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):22.4 - 24.7 Degree
- USP - pH:5.6 - 6.1
- USP - Loss on Drying at 105°C:≤0.3 %
- USP - Residue on Ignition:≤0.4 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Methionine sulfoxide:≤0.1 %
- USP - Related Compounds - N-Acetyl-D,L- methionine:≤0.2 %
- USP - Related Compounds - Any unspecified impurity:≤0.1 %
- USP - Related Compounds - Total unspecified impurities:≤0.30 %
- EP/BP - Assay (C₅H₁₁NO₂S) (dried basis):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:5.5 - 6.5
- EP/BP - Specific Rotation [Å]²º^D (+):22.5 - 24.0 Degree
- EP/BP - Related Substances: Unspecified Impurities, each:≤0.10 %
- EP/BP - Related Substances: Total Impurities:≤0.3 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):21.0 - 25.0 Degree
- JP - pH:5.2 - 6.2
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue after Ignition:≤0.10 %
- Endotoxin Concentration, IU/mg, For Information Only:
- Metallic Impurities - Cobalt (Co), For Information Only:
- Only Class 2 Solvents (Methanol) are likely to be present. All are:
- below Option 1 limits.:
- Elemental Impurities (USP (232), EP 5.20) - Information on:
- elemental impurities for this product is available on the:
- associated Product Regulatory Data Sheet and elemental impurity:
- profile report.:
Specification Test Results
About this item
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid,(S)-(-)-Methionine,L(-)-Methionine,(S)-(-)-2-Amino-4-(methylthio)butanoic acid,H-Met-OH,Methionine
Boiling Pt:278 °C (1013 hPa)
Melting Pt:136 °C
Density:1.373 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 278 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 136 °C
:Flash Pt:
:Flash Pt:
:M.W. 149.21 g/mol
Merck Index:13,06004
Storage Temperature:Ambient
EINECS:200-562-9
:M.O.C. Category R
Customers Who Bought This Also Bought