L(+)-Glutamic acid 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT2077-6
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 56-86-0
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:1 kg
- Cat. no.:CAJT2077-6
- Supplier No.:2077-06
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Assay (C₅H₉NO₄) (dried basis):98.5 - 100.5 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):30.5 - 32.5 Degree
- EP/BP - Ninhydrin-Positive Substances:Passes Test
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- USP - Assay (C₅H₉NO₄) (dried basis):98.5 - 101.5 %
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.02 %
- USP - Sulfate (SO₄):≤0.02 %
- USP - Iron (Fe):≤10 µg/g
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- USP - Specific Rotation [Å]²º^D (+):31.5 - 32.5 Degree
- USP - Loss on Drying at 105°C:≤0.1 %
- USP - Identification:Passes Test
- Endotoxin Concentration, EU/g:≤3.0
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
About this item
Synonyms: (S)-(+)-2-Aminopentanedioic acid,L(+)-1-Aminopropane-1,3-dicarboxylic acid,(S)-(+)-α-Glutamic acid,H-Glu-OH,L-Glutamic acid,L(+)-2-Aminopentanedioic acid,(S)-(+)-Glutamic acid,(S)-(+)-1-Aminopropane-1,3-dicarboxylic acid
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 325 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 160 °C
:Flash Pt:
:Flash Pt:
:M.W. 147.13 g/mol
Merck Index:13,04482
:L(+)-Glutamic acid
Storage Temperature:Ambient
EINECS:200-293-7
:M.O.C. Category R
Customers Who Bought This Also Bought