L-Arginine hydrochloride USP, Multi-Compendial, J.T.Baker®
Catalog #: CAJT2067-6
Supplier: Avantor
CAS Number: 1119-34-2
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:1 kg
- Supplier No.:2067-06
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- Meets J.P. Chemical Specifications:
- Preserve in well-closed containers.:
- Store in well-closed,light-resistant containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²º^D (+):21.4 - 23.6 Degree
- USP - Loss on Drying at 105°C:≤0.2 %
- USP - Residue on Ignition:≤0.1 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- USP - Chloride (Cl):16.5 - 17.1 %
- EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):21.0 - 23.5 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤0.5 %
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤0.02 %
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):21.5 - 23.5 Degree
- JP - pH (1 in 10):4.7 - 6.2
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.20 %
- JP - Residue after Ignition:≤0.10 %
- Endotoxin Concentration, IU/mg, For Information Only:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, L-Arginine hydrochloride USP, Multi-Compendial:
Specification Test Results
About this item
Synonyms: H-L-Arg-OH.HCl,(S)-2-Amino-5-guanidinopentanoic acid hydrochloride,(S)-(+)-Arginine HCl,L-Arginine, hydrochloride,(S)-(+)-Arginine hydrochloride,H-Arg-OH.HCl,L-Arg,L(+)-Arginine monohydrochloride
:Storage temperature: Ambient
Melting Pt:218 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 218 °C
:Flash Pt:
:Flash Pt:
:M.W. 210.66 g/mol
Storage Temperature:Ambient
EINECS:214-275-1
Customers Who Bought This Also Bought