ContactUsLinkComponent
extran* 300 Liquid Detergent for Manual Washing
Extran® 300 Liquid Detergent for Manual Washing, MilliporeSigma
Catalog #: CAEX0996-7
Supplier:  MilliporeSigma
Extran® 300 Liquid Detergent for Manual Washing, MilliporeSigma
Catalog #: CAEX0996-7
Supplier:  MilliporeSigma
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    20 L
  • Cat. no.:
    CAEX0996-7
Specifications
About this item
When diluted with water, concentrates can clean by themselves without leaving residues
Extran® 300 Liquid Detergent is a powerful liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues. Especially effective at removing organic materials such as blood and proteins. Phosphate- and halogen-free. Works well in ultrasonic cleaning baths. Can be used on glass, plastic, quartz, porcelain, ferrous metal, and rubber equipment and apparatus. Dose: 10–200mL per liter. pH of 1% solution: 10.1.
When diluted with water, concentrates can clean by themselves without leaving residues
Extran 300 Liquid Detergent is a powerful liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues. Especially effective at removing organic materials such as blood and proteins. Phosphate- and halogen-free. Works well in ultrasonic cleaning baths. Can be used on glass, plastic, quartz, porcelain, ferrous metal, and rubber equipment and apparatus. Dose 10 to 200 mL/L. pH of 1% solution is 10.1.
Extran 300 Liquid Detergent is a powerful liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues. Especially effective at removing organic materials such as blood and proteins. Phosphate- and halogen-free. Works well in ultrasonic cleaning baths. Can be used on glass, plastic, quartz, porcelain, ferrous metal, and rubber equipment and apparatus. Dose 10 to 200 mL/L. pH of 1% solution is 10.1.
For routine laboratory applications, no scrubbing is necessary once items are removed from the cleaning bath. For best results, items to be cleaned should be soaked for 2 to 24 hours, and rinsed with distilled water immediately after removal from cleaning bath.
1.07553.2500 is offered as an alternative to EX0994-2. Different Pack Size, Same Specifications.
1.07553.5000 is offered as an alternative to EX0994-4. Different Pack Size, Same Specifications.
1.07553.9025 is offered as an alternative to EX0994-7. Different Pack Size, Same Specifications.
1.07555.5000 is offered as an alternative to EX0993-4. different size, different packaging, same chem spec.
1.07555.9025 is offered as an alternative to EX0993-7. Different Pack Size, Same Specifications.
Extran® MA 03 Liquid Detergent is phosphate-free and safe for use on glass, plastic, quartz, porcelain, ferrous metals, and rubber equipment and apparatus. Well-suited for cleaning equipment used for the micro-determination of phosphate-based compounds. Dose: 20–200mL per liter. pH of 2% solution: 11.6.
Customers Who Bought This Also Bought