Corning® BioCoat® Collagen I-coated Plates, Corning
Catalog #: CACB356500
Supplier: Corning
Specifications
- Color:Clear
- Description:12-well, flat bottom TC-treated multiwell plate, with lid
- No. of wells:12
- Cat. no.:CACB356500
- Supplier No.:356500
Specifications
About this item
PS coated with rat tail collagen type I. Collagen is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adhesion, morphology, growth, migration, and differentiation of a variety of cell types. Applications include promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, studies of the effects of collagen I on cell behaviour, improving survival of primary cell lines in culture, and cell adhesion assays.
- Uniform application of rat tail collagen type I for consistent performance
- Manufactured in a highly controlled environment, and rigorously tested to assure product consistency and performance
Customers Who Bought This Also Bought