HOSE,NW25 FLEXIBLE 250MM S/S
Catalog #: CAC105-14-285
Supplier: Edwards
Customers Who Bought This Also Bought
Phenol ≥99% saturated solution in water, Biotechnology Grade, buffered
Supplier: VWR
$363.73 to $1,729.17
Glassine Weighing Paper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$24.71 to $27.82
Fused Quartz Microscope Slides and Coverslips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$45.79