Order Entry
ContactUsLinkComponent
30% BSA BOVINE ALBUMIN SOLUTION
Bovine Serum Albumin (BSA), Fraction V
Catalog #: CABSA-30
CAS Number:  
Bovine Serum Albumin (BSA), Fraction V
Catalog #: CABSA-30
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    Serum
  • Species:
    Bovine
  • Size:
    500 ml
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Concentration:
    30%
  • Cat. no.:
    CABSA-30
  • Origin:
    North America
  • Supplier No.:
    BSA-30

Specifications

About this item

The BSA is suitable for use in protease sensitive assays such as RIA, EIA, and nucleic acid hybridization. It is also used to remove non-specific binding or to stabilize proteins and enzymes. BSA, Fraction V is a 30% solution in 0.85% sodium chloride with 1% octanoic acid.

  • Preservative- and stabilizer-free.
Customers Who Bought This Also Bought