Order Entry
ContactUsLinkComponent
Ultra-Clear Centrifuge Tubes
Thinwall Ultra-Clear™ Tubes, Beckman Coulter
Catalog #: CABK344059
Supplier:  Beckman Coulter
Thinwall Ultra-Clear™ Tubes, Beckman Coulter
Catalog #: CABK344059
Supplier:  Beckman Coulter

Specifications

  • Dimensions:
    14×89 mm
  • Volume:
    13.2 ml
  • Cat. no.:
    CABK344059
  • Supplier No.:
    344059

Specifications

About this item

Thinwall Ultra-Clear™ tubes are for use in ultracentrifuge rotors.

  • Made of Ultra-Clear™ material

They have an open-top, thinwall closure type and are transparent, enabling easy location of visibly banded or pelleted samples.

Customers Who Bought This Also Bought