Falcon® 96-Well Library Storage Plates, Polypropylene, Corning
Catalog #: CABD353263
Supplier: Corning
Specifications
- Sterility:Non sterile
- Well Shape:V-Bottom
- Working Well Volume:250 µl
- Supplier No.:353263
Specifications
About this item
PP plate, PP lid, non sterile. Plates designed for compound storage.
- Rigid and flat plate for consistent automated handling
- Low retention PP and well design give low residual volumes after pipetting
- Lid compatible with automated lidding and de-lidding devices
- Raised well edges and flatness for heat and adhesive sealing
Customers Who Bought This Also Bought