ContactUsLinkComponent
Bulk Syringe, Nonsterile
Bulk Syringes, Nonsterile, BD Medical
Catalog #: CABD301030
Supplier:  BD
Bulk Syringe, Nonsterile
Bulk Syringes, Nonsterile, BD Medical
Catalog #: CABD301030
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    10 ml
  • Sterility:
    Non sterile
  • Tip Style:
    Slip
  • Supplier No.:
    301030
Specifications
About this item
Clean, ready-to-use, nonsterile syringes are siliconized and assembled with scale.
Syringes can be autoclaved or gas sterilized out of poly bags. Before autoclaving, remove plunger rod from barrel.
  • Syringes can be autoclaved or gas sterilized out of poly bags
  • Before autoclaving, remove plunger rod from barrel
Note: Syringes are supplied without needle. Needles must be purchased separately.
Ordering information: All syringes are bulk packed in double-wrapped poly bags. Sterile tip shield is sold separately.
Customers Who Bought This Also Bought