Protective Covers for NewClassic MS Series High Capacity Balances, METTLER TOLEDO®
Catalog #: CA97044-176
Supplier: Mettler Toledo
Specifications
- Description:Protective Cover for MS Balance 1 - 0.1 mg (Newclassic Ms-S Balances With A Draftshield)
- Cat. no.:CA97044-176
- Supplier No.:12121850
Specifications
About this item
These protective covers decreases the need to clean your balance and helps prevents cross contamination.
- Protects the balance against toxic or hazardous substances and provides operators with an increased level of safety
- The cover is installed and removed in a few simple steps. Once in place, it does not interfere with balance operation
Customers Who Bought This Also Bought