LAMP DEUTERIUM
Catalog #: CA97044-128
Supplier: Agilent Technologies
Customers Who Bought This Also Bought
Bromophenol blue 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$91.88 to $955.56
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Silver nitrate 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$297.60 to $1,622.45
Puritan® Miniature Polyester Tipped Applicator, Tapered Plastic Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
$422.35 to $4,223.39
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$120.43 to $1,204.30
Ammonium carbonate 30,0% (NH₃ basis), lumps, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$267.72 to $843.18
Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments
Supplier: Advanced Instruments
$748.29
EXEL International Brand Quality Luer Lock Syringes with Attached Needle, Air-Tite Products
Supplier: Air-Tite
$22.11 to $175.66
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
$186.99