ContactUsLinkComponent
Test Strip Hardness 50
Total Hardness Test Strips, Hach
Catalog #: CA97026-320
Supplier:  Hach
CAS Number:  
Total Hardness Test Strips, Hach
Catalog #: CA97026-320
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    CA97026-320
  • Range:
    0 - 425 mg/L
  • Supplier No.:
    2745250
  • No. of Tests:
    50
Specifications
About this item
Water hardness is caused almost entirely by calcium and magnesium ions. Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers.
Easy to use, disposable, and inexpensive.
Easy to use, disposable, and inexpensive.
A great way to obtain quick, quantitative answers in the field or in the lab.
Customers Who Bought This Also Bought