Order Entry
ContactUsLinkComponent
PANS ALUM 40 UL 0.15 MM PK400
Aluminium Sample Pans, PerkinElmer
Catalog #: CA97012-266
PANS ALUM 40 UL 0.15 MM  PK400
Aluminium Sample Pans, PerkinElmer
Catalog #: CA97012-266

Specifications

  • Volume:
    40 µl
  • Description:
    Aluminum Pans without Covers
  • Supplier No.:
    B0143021

Specifications

About this item

Each sample pan and cover has an operating range of -170° to 600 °C, and an ambient max pressure.

  • Can be used with nonvolatile solid samples
  • Crimped design
  • Not hermetically sealed

These sample pans can be used with nonvolatile solid samples, such as plastics and polymers, and are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped, but not hermetically sealed. Not for use with Diamond DSC, PYRIS 1 DSC, or DSC 7 Autosamplers.

Customers Who Bought This Also Bought