Order Entry
ContactUsLinkComponent
CASTER 5 STM W/BPR
MetroMax i™/MetroMax iQ™ Stem Casters, Metro™
Catalog #: CA97005-752
Supplier:  Metropolitan Wire
CASTER 5 STM W/BPR
MetroMax i™/MetroMax iQ™ Stem Casters, Metro™
Catalog #: CA97005-752
Supplier:  Metropolitan Wire

Specifications

  • Load Capacity:
    300 lbs.
  • Description:
    Polyurethane Swivel Caster with All-Polymer Horn; Microban antimicrobial product protection
  • Cat. no.:
    CA97005-752
  • Supplier No.:
    5PCXM

Specifications

About this item

Use with MetroMax i™ or MetroMax iQ™ shelving to create mobile units.

  • Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
  • High modulus rubber casters provide extra shock protection
  • Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbent

Casters with all-polymer horn offer corrosion resistance and enhanced durability.

Each caster includes a protective bumper. Casters feature a tread width of 3.2 cm (⁵/₄") and a wheel diameter of 12.7 cm (5"). Swivel, foot-operated brake, and rigid styles are available.

Ordering information: Rigid casters are held in place by a connecting channel; when ordering rigid casters, shelf depth must be provided.
Customers Who Bought This Also Bought