Order Entry
ContactUsLinkComponent
Dna Binding Plate
E-Z 96 DNA Plates, Omega Bio-tek
Catalog #: CA95030-226
Supplier:  Omega Bio-Tek
CAS Number:  
E-Z 96 DNA Plates, Omega Bio-tek
Catalog #: CA95030-226
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    E-Z 96 DNA Plates
  • Size:
    10 plates
  • Description:
    E-Z 96 DNA Plates
  • Cat. no.:
    CA95030-226
  • Supplier No.:
    BD96-01

Specifications

About this item

E-Z 96 plates are silica glass fiber plates that can bind up to 50 μg of genomic DNA or 20 μg of plasmid DNA per prep

Each well has a capacity of 1 mL and the plates are semi skirted. E-Z 96 DNA plates cab be process in vacuum manifolds and/or by centrifugation. Omega Bio-tek Columns are compatible with following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, Dneasy Plant DNA.

Customers Who Bought This Also Bought