To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Donkey
- Isotype:IgG
- Reactivity:Rabbit
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- Size:100 µL
- Western blot:Yes
- Antibody fragment:Whole Ab
- Storage buffer:Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
- Storage temperature:2 - 8 °C
- Immunogen:Rabbit IgG
- Cat. no.:CA95017-328L
- Supplier No.:NA934-100UL
Specifications
About this item
Highly species specific HRP-conjugated antibody optimized for use with Amersham ECL Western Blotting Detection Reagents.
Optimized for use with our range of Amersham ECL Western blotting detection reagents.
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2°C to 8°C.
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit