Order Entry
ContactUsLinkComponent
DNA Starter Kit Flashgel
FlashGel™ DNA, RNA and Recovery Systems
Catalog #: CA95015-612
Supplier:  Lonza
DNA Starter Kit Flashgel
FlashGel™ DNA, RNA and Recovery Systems
Catalog #: CA95015-612
Supplier:  Lonza
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    FlashGel DNA Starter Kit
  • Cat. no.:
    CA95015-612
  • Includes:
    Pack of 1.2% DNA Cassettes, Loading Dye, DNA Marker and Dock
  • Supplier No.:
    57026

Specifications

About this item

FlashGel Systems consist of enclosed, disposable, precast agarose gel cassettes and a combination electrophoresis and transilluminator unit.

  • Systems provide nearly instant results
  • Simply load samples, watch bands migrate and get data in as little as two minutes

FlashGel DNA system separates DNA in 2–7 minutes with outstanding resolution and allows monitoring of gel runs right at the bench without UV light

FlashGel RNA system is a fast and sensitive tool for analysis of sample integrity and optimized for the unique requirements of RNA. Recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity. The system provides results in 30 minutes or less while avoiding use of hazardous reagents and contaminating RNases. RNA fragments are analyzed on the FlashGel® system using the same fast and simple procedure used for DNA. Separation is complete in less than eight minutes and RNA samples are ready for imaging within 10 to 20 minutes. FlashGel® system for RNA is 5 to 20 times more sensitive than ethidium bromide gels and RNA <10 ng per band are clearly detected, conserving precious RNA samples.

FlashGel system for DNA recovery eliminates agarose gel preparation and delivers highly efficient recovery, free from inhibitors and UV-induced damage, in a simple 5 to 10 minute protocol. Go from sample loading to recovery in just five minutes. Recover samples directly from the cassette without band excision or purification. Visualize sample recovery without UV light and recover at 80 to 100% efficiency.

FlashGel Dock is an electrophoresis apparatus with a built-in transilluminator that provides visualization of both DNA and RNA cassettes. The FlashGel camera captures gel images from the FlashGel system right at the benchtop with complete gel run and image capture in just 5 minutes. Camera software is compatible with PC computers.

The highly sensitive system also allows 5 to 20 times the reduction in DNA levels, saving both time and money.

Customers Who Bought This Also Bought