Water, Cell Culture Grade, tested to USP specifications for injection, sterile, Corning®
Catalog #: CA95000-092
Supplier: Corning
CAS Number: 7732-18-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Size:6× 100 ml
- Cat. no.:CA95000-092
- Supplier No.:25-055-CI
Specifications
Specification Test Results
- Mycoplasma:Tested negative
- Plating efficiency:Pass
- Oxidizable substances Solution:remains faintly pink
- Total organic carbon (before packaging) USP <643>:< 500 ppb
- Water conductivity (packaged water) USP <645>:Meets USP Requirements
- Water conductivity (before packaging) USP <645>:Meets USP temperature/conductivity requirements
- Endotoxin USP <85>:≤ 0.025 EU/mL
- Sterility USP <71>:Pass
- Particulate matter USP <788>:≤ 25 particles/mL that are ≥ 10 μm in size
≤ 3 particles/mL that are ≥ 25 μm in size - Corning®, Water, Cell Culture Grade, tested to USP specifications for injection, sterile:
Specification Test Results
About this item
Synonyms: Hydrogen oxide
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:Water
:0
:100
Storage Temperature:Ambient
:12,10175
Customers Who Bought This Also Bought