Order Entry
ContactUsLinkComponent
GRAM CRYSTAL VIOLET SOLUTION 4 L
BD BBL™ and BD Difco™ Gram Stain Kit Components, BD Diagnostics
Catalog #: CA90008-896
Supplier:  BD
CAS Number:  
GRAM CRYSTAL VIOLET SOLUTION 4 L
BD BBL™ and BD Difco™ Gram Stain Kit Components, BD Diagnostics
Catalog #: CA90008-896
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Gram Crystal Violet
  • Size:
    3.8 L (1 gal.)
  • Cat. no.:
    CA90008-896
  • Supplier No.:
    212526

Specifications

About this item

Each Gram crystal violet solution contains approximately 0.3% crystal violet aqueous alcohol solution

Gram stain QC slides are conventional 25x75mm (1x3") microscope slides imprinted with ten squares. One square contains gram-positive and gram-negative control organisms Staphylococcus aureus and E. coli and the remaining nine squares are available for staining test isolates.

Each Gram decolorizer solution contains a 3:1 ratio of ethanol to acetone to assure even and consistent decolorization. Each Gram iodine solution (stabilized) contains an organically stabilized iodine-polyvinylpyrrolidone (PVP), which retains a safe and constant level of unoxidized iodine to maintain effectiveness as a mordant for at least 15 months. Gram iodine (unstabilized) is designed to prepare a traditional Gram’s working solution without requiring a breakage of ampules. Safranin is a mixture of dimethyl and trimethyl and is most commonly used for counterstaining nuclei red. Basic Fuchsin is a mixture of Pararosanilin, Rosanilin, and Magenta II.

Ordering information: All containers feature no-drip cap spout. Kits are packaged in convenient compact trays.
Customers Who Bought This Also Bought