ContactUsLinkComponent
Nitrate Broth
Difco™ Nitrate Broth, BD Life Sciences
Catalog #: CA90001-916
Supplier:  BD
CAS Number:  
Nitrate Broth
Difco™ Nitrate Broth, BD Life Sciences
Catalog #: CA90001-916
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Nitrate Broth
  • Size:
    500 g
  • Cat. no.:
    CA90001-916
  • Supplier No.:
    226810
Specifications
About this item
Nitrate Broth is recommended as an aid in the identification of aerobic and facultative anaerobic gram-negative microorganisms by means of the nitrate reduction test.
  • Basal medium containing potassium nitrate
  • Addition of zinc dust is used to detect unreduced nitrate
  • Organisms derive oxygen from nitrate through anaerobic respiration
The formation of nitrogen gas, an end product typical of certain organisms, is evidenced by displacement of the medium from the Durham tube by the gas produced.
Customers Who Bought This Also Bought