ContactUsLinkComponent
LOOP INOC LT BLU 10UL PK1000
Disposable Inoculating Loops and Needles, Sterile, BD Difco™
Catalog #: CA90001-102
Supplier:  BD
LOOP INOC LT BLU 10UL PK1000
Disposable Inoculating Loops and Needles, Sterile, BD Difco™
Catalog #: CA90001-102
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Blue
  • Description:
    Inoculating Loops
  • Size:
    10 μL
  • Packaging:
    50 per Tube
  • Supplier No.:
    220217
Specifications
About this item
Made of specially formulated plastic resin
Flexible enough to prevent gouging agar yet firm enough for picking difficult colonies. Comfortable, smooth-textured, flat handle. Color-coded for easy size identification.
Ordering information: Packaged sterile, ready to use, in tamper-evident, resealable clear plastic tubes.
Customers Who Bought This Also Bought