ContactUsLinkComponent
BD BBL* CultureSwab* Liquid Stuart, Single Swab
BD BBL™ Microbiological Specimen Collection and Transport Systems, BD Diagnostics
Catalog #: CA90001-040
Supplier:  BD
BD BBL* CultureSwab* Liquid Stuart, Single Swab
BD BBL™ Microbiological Specimen Collection and Transport Systems, BD Diagnostics
Catalog #: CA90001-040
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Liquid Stuart, Single Swab
  • Cat. no.:
    CA90001-040
  • Application:
    For vaginal, skin, wound, and throat specimens
  • Supplier No.:
    220099
Specifications
About this item
Keep samples viable during transport with convenient, self-contained BD CultureSwab™ and BD CultureSwab Plus™ systems
Each BD CultureSwab™ tube contains a swab with a polyurethane sponge. After sampling, the swab is returned to the tube and specimen is put in immediate contact with the medium and kept moist during transport.
Each BD CultureSwab Plus™ tube contains a 5mL column of Amies gel for maximum specimen protection and improved sample viability of anaerobes. Gel column also ensures that swab maintains a centralized position during transport. BD CultureSwab Plus™ tubes feature a special venturi hourglass design that helps prevent breakdown of the gel column during transport, minimizes bubbles and breaks in gel, and reduces the formation of air pockets that are harmful to fastidious bacteria.
Both are ideal for sampling aerobes; BD CultureSwab Plus™ is also appropriate for strict anaerobes. Systems consist of tube, transport medium, and swab with integral cap. Tubes are made of unbreakable polypropylene and include a preattached data label and a tamper-evident seal. Swabs are rayon and are designed with an extra long shaft. Various media are available for vaginal, skin, wound, nasopharyngeal, ear, nose, throat, and eye specimens, as well as male urethral sampling. Systems are also available with Cary-Blair agar for fecal specimens and with a sterile swab in an empty tube for general specimen collection and transport. Each swab package is flushed with nitrogen gas to assure freshness of the medium throughout the product shelf life.
Customers Who Bought This Also Bought