Order Entry
ContactUsLinkComponent
Agar, Bacto*
Bacto™ Agar Soldifying Agent, BD Diagnostics
Catalog #: CA90000-760
Supplier:  BD
CAS Number:  
Agar, Bacto*
Bacto™ Agar Soldifying Agent, BD Diagnostics
Catalog #: CA90000-760
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Agar, BD Bacto
  • Size:
    454 g
  • Cat. no.:
    CA90000-760
  • Supplier No.:
    214010

Specifications

About this item

Bacto Agar is recommended for clinical applications, auxotrophic studies, bacterial and yeast transformation studies and bacterial molecular genetics applications.

  • Recommended for clinical applications
  • Solidifying agent
  • Optimized for beneficial calcium and magnesium content

A solidifying agent, is a purified agar in which extraneous matter, pigmented portions and salts are reduced to a minimum

Optimized for beneficial calcium and magnesium content, and reduction of detrimental ions such as copper and iron. It is available in the form of fine, light colored granules which are convenient for weighing and handling.

It is usually used in concentrations of 1 - 2% in solid culture media or in concentrations of 0.05% - 0.5% in media used for determination of motility and the growth of anaerobes and microaerophile.

Customers Who Bought This Also Bought