ContactUsLinkComponent
Potato Dextrose Agar
Difco™ Potato Dextrose Media, BD
Catalog #: CA90000-758
Supplier:  BD
CAS Number:  
Difco™ Potato Dextrose Media, BD
Catalog #: CA90000-758
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Potato Dextrose Agar
  • Size:
    500 g
  • Cat. no.:
    CA90000-758
  • Supplier No.:
    213400
Specifications
About this item
Potato Dextrose Agar is used for the cultivation and enumeration of yeasts and molds.
  • Potato starch and dextrose support luxuriant growth of fungi
  • Lowering the pH of the medium inhibits bacterial growth
  • Recommended media for testing nonsterile pharmaceutical products
Potato Dextrose Agar is a general-purpose medium for yeasts and molds that can be supplemented with acid or antibiotics to inhibit bacterial growth.
Potato Dextrose Broth is a general-purpose broth medium for cultivating yeasts and molds.
Customers Who Bought This Also Bought