ContactUsLinkComponent
Oxgall
Difco™ Oxgall, BD Diagnostics
Catalog #: CA90000-728
Supplier:  BD
CAS Number:  
Difco™ Oxgall, BD Diagnostics
Catalog #: CA90000-728
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Oxgall
  • Size:
    500 g
  • Cat. no.:
    CA90000-728
  • Supplier No.:
    212820
Specifications
About this item
Dehydrated fresh bile used in the selection of bile-tolerant bacteria.
  • Inhibits the growth of gram-positive microorganisms
  • Provides a regular supply of bile
  • Used to aid the determination of enteric pathogens
Oxgall is manufactured from large quantities of fresh bile by rapid evaporation of the water content. Bile is composed of fatty acids, bile acids, inorganic salts, sulfates, bile pigments, cholesterol, mucin, lecithin, glycuronicacids, porphyrins, and urea.
Customers Who Bought This Also Bought