ContactUsLinkComponent
BBL* Mycoplasma Broth Base
Mycoplasma Broth Base, BD Diagnostics
Catalog #: CA90000-200
Supplier:  BD
CAS Number:  
BBL* Mycoplasma Broth Base
Mycoplasma Broth Base, BD Diagnostics
Catalog #: CA90000-200
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mycoplasma Broth Base, without Crystal Violet (CV)
  • Size:
    500 g
  • Cat. no.:
    CA90000-200
  • Supplier No.:
    211458
Specifications
About this item
Mycoplasma broths, when supplemented with nutritive enrichments, are used for isolating and cultivating Mycoplasma.
  • Peptones and beef extract provide essential source of nitrogen
  • Yeast extract is a source of vitamins and carbon
  • Sodium chloride maintains the osmotic balance
Mycroplasma Broth Base without crystal violet is prepared according to the formula described by Morton and Lecci. Crystal violet is omitted from this formula due to its inhibitory action on some Mycoplasma. It has been used for the cultivation of Mycoplasma for research studies.
Customers Who Bought This Also Bought