ContactUsLinkComponent
MacConkey Agar
MacConkey Agar, BD
Catalog #: CA90000-168
Supplier:  BD
CAS Number:  
MacConkey Agar
MacConkey Agar, BD
Catalog #: CA90000-168
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MacConkey Agar
  • Size:
    500 g
  • Cat. no.:
    CA90000-168
  • Supplier No.:
    211387
Specifications
About this item
MacConkey agar is a selective and differential culture medium for bacteria designed to selectively isolate Gram-negative and enteric bacilli and differentiate them based on lactose fermentation.
  • Isolates Gram-negative bacilli
Enteric bacteria that have the ability to ferment lactose can be detected using the carbohydrate lactose, and the pH indicator neutral red.
Customers Who Bought This Also Bought