Matrix Filtered D.A.R.T.'s Tips, Thermo Scientific
Catalog #: CA89168-960
Supplier: Thermo Fisher Scientific
Specifications
- Volume:300 µl
- Sterile:+
- Tip Style:96 Tips, Extended Length
- Supplier No.:5538
Specifications
About this item
Unlike conventional automation tips, Matrix D.A.R.T.s tips are provided in disposable, rigid magazines that bond onto the pipetting heads by sealing against a silicone pad.
Compatible with the Versette, Hydra DT, PlateMate 2x3 and PlateMate Plus Automated Pipetting Systems. Stepped-body design minimizes volume displacement and provides tip straightness for better well access.
This forms a definitive seal that minimizes tip-to-tip height variations and eliminates static interactions, delivering.
Customers Who Bought This Also Bought