Matrix D.A.R.T.'s Tips, Thermo Scientific
Catalog #: CA89168-938
Supplier: Thermo Fisher Scientific
Specifications
- Volume:30 µl
- Tip Style:96 Tips, Extended Length
- Supplier No.:5506
Specifications
About this item
Unlike conventional automation tips, Matrix D.A.R.T.s tips are provided in disposable, rigid magazines that bond onto the pipetting heads by sealing against a silicone pad.
- Disposable, rigid magazines
- Definitive seal minimizes tip-to-tip height variations
- Consistent dispensing into the bottom of microplate wells
Compatible with the Versette, Hydra DT, PlateMate 2×3 and PlateMate Plus Automated Pipetting Systems. Stepped-body design minimizes volume displacement and provides tip straightness for better well access. Disposable, rigid magazines form a definitive seal that minimizes tip-to-tip height variations and eliminates static interactions, delivering consistent dispensing into the bottom of microplate wells.
Customers Who Bought This Also Bought