Automation Pipette Tips for PlateMate, PlateMate Plus, and Versette Workstations, Thermo Scientific
Catalog #: CA89168-926
Supplier: Thermo Fisher Scientific
Specifications
- Volume:100 µL
- Tip Color:Clear
- Packed:96 Tips/Rack, 20 Racks/Case
- Cat. no.:CA89168-926
- Tip Style:Standard
- For Use With:Thermo Scientific automated workstations
- Supplier No.:5326
Specifications
About this item
Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot.
Each lot must pass a 15-point QC inspection including performance testing on our PlateMate workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Customers Who Bought This Also Bought