ContactUsLinkComponent
Heat Sealing PCR Foil
PCR Films and Foils
Catalog #: CA89167-286
Supplier:  Eppendorf
Heat Sealing PCR Foil
PCR Films and Foils
Catalog #: CA89167-286
Supplier:  Eppendorf
Specifications
  • Description:
    Heat sealing foil; Long-term storage; protection against evaporation during PCR; easily pierced by Pipette tips; can be removed from plate
  • Supplier No.:
    0030127854
Specifications
About this item
Ensure airtight sealing of your PCR plates with Eppendorf® PCR films and foils. Tailor your choice to your assay needs, like the Masterclear® real-time PCR film, optimal for qPCR with its superior optical characteristics, or heat-sealing options for the best evaportion protection.
  • PCR plate seals offer excellent chemical resistance and have been tested for resistance to a variety of common laboratory substances
  • Specialized seals with optimal optical properties are available to help minimize signal loss in qPCR
Eppendorf PCR films and foils provide effective and airtight sealing for plates to help protect PCR samples during thermal cycling and storage.
Customers Who Bought This Also Bought