Specifications
- Clothing Size:XL/XXL
- Garment application:Cleanroom,Laboratory
- Color:White
- Description:Boot covers
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA89166-202
- Supplier No.:12919
Specifications
About this item
KIMTECH PURE® A5 Clean Processed apparel offers a high-value combination of comfort and quality, and incoporates innovative design features to provide an exceptionally high standard of protection.
- High standard of protection and comfort
- Cloth-like SMS material, contains no natural rubber or latex
- Manufactured in ISO-certified facility, individually bagged under cleanroom conditions
- Optional hoods and boot covers designed for use with coveralls
Cloth-like comfort is delivered by the classic Kimberly-Clark SMS (Spunbond Meltblown Spunbond) material that goes into the product
All coveralls features patented REFLEX™ design, which is 7.5 times more rip-resisitant than ANSI minimums. Built-in thumb loops keep sleeves from rolling back and exposing the wearer. Coveralls feature a mandarin collar that rises higher on the neck for added protection. All apparel is equipped with tunneled elastic and bound seams. Triple bagged. All items are suitable for ISO Class 5 or higher cleanrooms.
These garments are manufactured in an ISO certified facility, clean processed, and individually bagged under cleanroom conditions. They contain no natural or rubber latex. The optional hoods and boot covers are designed for use with the coveralls. This A5 apparel is ideally suited for ISO 5–8 Cleanroom environments in pharmaceutical, biotech, and medical device industries. It is easy to don, comfortable to wear, and efficient to stock.