Nutra-Gel Diet™ Purified Formula, Rodent, Sterile, Bio-Serv
Catalog #: CA89132-274
Supplier: Bio-Serv
Specifications
- Description:Nutra-Gel Tray, Bacon Flavor
- Size:40 g/cube
- Cat. no.:CA89132-274
- Supplier No.:S5769-TRAY
Specifications
About this item
Nutritionally complete, purified, bacon-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, weanlings, or transportation
- Trays contain 16 pre-measured cubes that will pop out as needed and can be placed directly on the bedding or in a food hopper
- Nutra-Gel remains moist in the cage 24 to 48 hours, depending on the type of cage system
- Convenient and ready to serve; no mixing required
- Each cell is individually sealed, which maintains moisture and sterility
- Phytoestrogen free
- Bacon flavor stimulates appetite
- Nutritionally assayed
- Shelf Life: 6 months frozen, 3 months refrigerated
Ingredients: Water, Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ.
Caloric value: 1.57 kcal/g
Nutritional ash content: 0.9%
Nutritional carbohydrate content: 22.2%
Nutritional fat content: 3.3%
Nutritional fibre content: 1.9%
Nutritional moisture content: <70%
Nutritional protein content: 9.5%
Ordering information: Most mice consume approximately 10-12 g of Nutra-Gel per day. Rats consume approximately 26-60 g of Nutra-Gel per day, depending on strain and age.
Customers Who Bought This Also Bought