INTERFERin®, siRNA and miRNA transfection reagent, Polyplus-transfection
Catalog #: CA89129-928
Supplier: Polyplus Transfection
CAS Number:
Specifications
- siRNA:Yes
- Description:INTERFERin®, siRNA and miRNA transfection reagent
- Size:100 µl
- No. of Reactions:50 - 100 (24 wells)
- Supplier No.:409-01
Specifications
About this item
Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
INTERFERin® is a powerful, ready to use siRNA and miRNA reagent which provides more than 90% silencing efficiency at 1 nM siRNA in a wide variety of cells.
- Great silencing using as little as 1 nM siRNA
- Over 90% gene silencing in a wide variety of cells
- Suitable for transfection miRNA and other oligonucleotides
- Gentle mode of action for more robust data and excellent cell viability
- Compatible with serum and antibiotics
Using low concentration of siRNA avoids off-target effects, and its gentle mode of action ensure more robust data and excellent cell viability
Easy to use thanks to its compatibility with serum and antibiotics, INTERFERin® is also perfectly suited for transfection of miRNA and other oligonucleotides such as pre-miRNA, mimic miRNA, antimiR, etc...
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of INTERFERin® is tested in house in a transfection assay on A549-Luc cells, constitutively expressing the Luciferase gene. The silencing efficiency obtained using 1 nM siRNA and INTERFERin® for each batch is indicated on the Certificate of Analysis.
Every batch of INTERFERin® is tested in house in a transfection assay on A549-Luc cells, constitutively expressing the Luciferase gene. The silencing efficiency obtained using 1 nM siRNA and INTERFERin® for each batch is indicated on the Certificate of Analysis.
Delivery: 1 mL of INTERFERin® is sufficient to perform 500 to 1000 transfections in 24-well plates.
Customers Who Bought This Also Bought