jetPRIME®, DNA and siRNA Transfection Reagent, Polyplus-transfection
Catalog #: CA89129-924
Supplier: Polyplus Transfection
CAS Number:
Specifications
- CRISPR/Cas 9:Yes
- DNA:Yes
- DNA and siRNA:Yes
- siRNA:Yes
- Virus production:Yes
- Description:jetPRIME®, DNA and siRNA transfection reagent
- Size:1.5 ml
- Cat. no.:CA89129-924
- Buffer Volume:2×60 ml
- No. of Reactions:375
- Supplier No.:114-15
Specifications
About this item
jetPRIME® DNA and/or siRNA transfection reagent is a powerful, and versatile reagent designed to ensure high DNA transfection efficiency and excellent gene silencing in a variety of adherent cells.
- High DNA transfection efficiency
- Low amounts of nucleic acid
- Superior cell viability
- One reagent for DNA and/or siRNA transfections
- Cost-effective
jetPRIME® is ideal for co-transfection of DNA and siRNA. jetPRIME® is gentle to cells since it requires low amounts of reagent and nucleic acid during transfection. Effective and non toxic DNA and siRNA delivery is essential for reliable scientific results.
375 transfections in 6-well plates.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Delivery: Free of charge samples are available upon request. Please contact your local Avantor sales office for further details. Supplied with an optimised sterile buffer which must be used to ensure successful transfection experiments. The concentrated buffer provided with 114-75C must be diluted 1:5 in sterile H₂O just prior to use. 1.5 ml is sufficient to perform.
Customers Who Bought This Also Bought