jetPEI®, HTS DNA Transfection Reagent
Catalog #: CA89129-916
Supplier: Polyplus Transfection
CAS Number:
Specifications
- DNA:Yes
- High throughput screening:Yes
- Description:jetPEI®, HTS DNA transfection reagent
- Size:1 ml
- Buffer Volume:50 ml
- No. of Reactions:1200 - 1600
- Supplier No.:101-10N
Specifications
About this item
jetPEI® transfection reagent is a linear polyethylenimine derivative, free of components of animal origin, providing highly effective and reproducible gene delivery, perfectly suited for High Throughput Screening (HTS) applications.
- Well-suited for adherent and suspension cells
- Fast and efficient methods to transfect cells for HTS
- Compatible with serum and antibiotics
- Exceptionally reproducible results
- Reverse, batch & forward protocols available
jetPEI® transfection reagent is particularly well suited for automated or manual High Throughput Screening (HTS) with three protocols available: Reverse, batch and forward.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of jetPEI® is tested in-house by DNA transfection of HeLa cells. Transfection with a firefly Luciferase gene under the control of CMV promoter gives at least 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Every batch of jetPEI® is tested in-house by DNA transfection of HeLa cells. Transfection with a firefly Luciferase gene under the control of CMV promoter gives at least 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Delivery: 150 mM NaCl solution is included. 1 mL of jetPEI® transfection reagent is sufficient to perform 2000 transfections in 96-well plates
Customers Who Bought This Also Bought