DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood, Boots, and Thumb Loops
Catalog #: CA89129-596
Supplier: DuPont
Specifications
- Clothing Size:2XL
- Garment application:Cleanroom,Laboratory
- Color:White
- Cat. no.:CA89129-596
- Chest Size:283/4"
- Inseam:301/2"
- Supplier No.:IC105SWH2X0C
- Sleeve:381/4"
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Serged seams
- Set sleeve design
- Elastic wrists and ankles
- Attached thumb loops
- Zipper closure
- Attached boots with PVC sole
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Thumb loops minimize bunching sleeves. Attached hood with elastic around face opening ensures a secure fit. Front zipper closure allows for easy donning and doffing. Soles provide enhanced skid-resistance and durability.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners. Clean Processed and Clean Processed, Sterile garments are individually packaged.
Customers Who Bought This Also Bought