Specifications
- Garment application:Cleanroom
- Length:151/4"
- Shoe size:L
- Color:White
- Height:18"
- Cat. no.:CA89127-324
- Supplier No.:IC457SWHLG0S
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
Clean-processed garments are processed to minimize particle shedding and individually packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Sterile garments are gamma sterilized to a Sterility Assurance Level (SAL) of 10-6.
- PVC sole
- Elastic opening
- Ankle ties
- Coverage up to calf
- Serged seams
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Ankle ties allow for a secure fit. PVC soles provide enhanced skid-resistance and durability.