Eppendorf® DNA LoBind Plates
Product Details & Documents
DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
- Eppendorf LoBind material ensures optimized sample recovery for improved assay results
- Free of surface coating (e.g. silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors
- RecoverMax® well design: Optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface ensure reliable sealing
Maximize recovery of nucleic acids can be achieved with DNA LoBind® plates. A combination of special manufacturing technologies and selected polypropylene batches ensure nearly 100 % recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination. This makes Eppendorf LoBind Plates ideal for applications where sample recovery is key - such as the preparation of long-term storage nucleic acids in forensic, microarrays and NGS applications.
Purity grade: PCR Clean
Material: Polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Well color: Clear
Specifications
- Well volume:500 µl
- Color:White
- Description:Deepwell plate
- Packaging:40 plates (5 bags × 8 plates)
- Cat. No.:CA89085-650
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



