Dustless Precision Pellets® Rodent Grain-Based Diet, Bio-Serv
Catalog #: CA89067-546
Supplier: Bio-Serv
Specifications
- Cat. no.:CA89067-546
- Packaging:50,000/Box
- Weight:20 mg
- Supplier No.:F0163
Specifications
About this item
Precise size and weight pellets generally used in nutritional and behavioral studies.
- Dustless and fracture free
- Superior performance in automatic feeders
- Nutritionally complete, banana-flavored, grain-based formula
- Available in purified formula
- Wide range of weights: 20 mg to 1 g
- Every batch tested for hardness, weight and height
- Custom flavors and colors available upon request
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.
Caloric value: 3.35 kcal/g
Nutritional ash content: 8.1%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 3.8%
Nutritional fibre content: 4.0%
Nutritional moisture content:<10%
Nutritional protein content: 21.3%
Customers Who Bought This Also Bought