Doxycycline Rodent Diet, Sterile, Bio-Serv
Catalog #: CA89067-462
Supplier: Bio-Serv
Specifications
- Description:Doxycycline 200 mg/kg, Sterile
- Size:Pellets
- Cat. no.:CA89067-462
- Packaging:10 kg/Box
- Supplier No.:S3888
Specifications
About this item
The easiest, most precise research tool to manipulate doxycycline sensitive transgenes.
- Doxycycline in a pelleted form is more stable than in water
- Nutritionally complete grain-based rodent diet containing 200 mg of doxycycline per kg of diet (most common concentration)
- The light green color differentiates Dox Diet from standard feed
- Sterilized by gamma irradiation. Doxycycline Diet can be customized to meet the exact concentration needed
- Nutritionally assayed
- Available non-sterile
- Shelf Life: 6 months, cool dry storage
Ingredients: Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer's Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride, Doxycycline, Food Dye.
Caloric value: 3.75 kcal/g
Nutritional ash content: 4.8%
Nutritional carbohydrate content: 53.6%
Nutritional fat content: 8.7%
Nutritional fibre content: 2.1%
Nutritional moisture content: <10%
Nutritional protein content: 20.8%
Customers Who Bought This Also Bought